missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human NEK2, His Tag Recombinant Protein

Product Code. 11573437
Click to view available options
Quantity:
1 mg
10 μg
Unit Size:
10µg
1mg
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11573437

Brand: Invitrogen™ PV3360

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length.

NEK2 is involved in cell cycle regulation and is essential for faithful chromosome segregation. NEK2 is a gene that encodes for a serine/threonine-protein kinase. This protein plays a crucial role in the regulation of mitosis and is primarily located in the centrosome. During the G1 phase, the protein is undetectable, but its levels progressively increase throughout the S phase, reaching their peak in late G2 phase. The gene has several alternatively spliced transcript variants that encode different isoforms with distinct C-termini. NEK2 is associated with several diseases, including Retinitis Pigmentosa 67 and Retinitis Pigmentosa.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P51955
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris with 500mM NaCl, 0.5mM EDTA, 0.05% Triton X-100, 50% glycerol, 2mM DTT and no preservative; pH 7.5
Gene ID (Entrez) 4751
Molecular Weight (g/mol) 54.5 kDa
Name Human NEK2, His Tag
pH Range 7.5
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias AA617254; C77054; HSPK 21; HsPK21; NEK 2; NEK 2A; Nek2; Nek2 kinase; NEK2A; Never in mitosis A-related kinase 2; NIMA - related expressed kinase 2; NIMA (never in mitosis gene a)-related expressed kinase 2; NIMA (never in mitosis gene a)-related kinase 2; NIMA related kinase 2; nimA-like protein kinase 1; NIMA-related expressed kinase 2; NIMA-related kinase 2; nimA-related protein kinase 2; NLK 1; NLK1; PPP1R111; protein phosphatase 1, regulatory subunit 111; RP67; Serine/threonine-protein kinase Nek2
Common Name NEK2
Gene Symbol NEK2
KinaseFamily NEK Kinase Family
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Assay Entry Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical binding/kinetic
Kinase Group Ser⁄Thr Kinases: Other
Shipping Condition Dry Ice
System Compatibility LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™
Target Entry NEK2
Target Gene NIMA (never in mitosis gene a)-related kinase 2
Expression System Baculovirus
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Full Length
Form Liquid
Protein Subtype Serine/Threonine Kinases
Research Category Signal Transduction
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.