Learn More
Eurofins DiscoverX NEK2 Protein, Active
Description
NEK (NIMA-Related Kinase) Ser/Thr kinase family includes at least 11 members and is involved in regulation of mitosis. Overexpression of active NEK2 (NLK1) induces a striking splitting of centrosomes, whereas prolonged expression of either active or inactive NEK2 leads to dispersal of centrosomal material and loss of a focused microtubule-nucleating activity.
GenBank NM_002497
Specifications
Specifications
| Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Molecular Weight (g/mol) | 52.8 kDa |
| Quantity | 10 μg |
| Species | Human |
| Recombinant | Recombinant |
| Protein Tag | N-terminal 6His |
| Expression System | Recombinant enzyme expressed in Sf21 insect cells |
| Protein Form | Full-length |
| Purity or Quality Grade | ≥49% |
| Protein | NEK2, active |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.