missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NEK10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA581225
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human NEK10.
NEK10 (NIMA Related Kinase 10) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene includetransferase activity, transferring phosphorus-containing groupsandprotein tyrosine kinase activity.
Specifications
| NEK10 | |
| Polyclonal | |
| Unconjugated | |
| NEK10 | |
| FLJ32685; NEK10; Never in mitosis A-related kinase 10; NIMA (never in mitosis gene a)- related kinase 10; NIMA related kinase 10; NIMA-related kinase 10; nimA-related protein kinase 10; Serine/threonine-protein kinase Nek10 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 152110 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q6ZWH5 | |
| NEK10 | |
| A synthetic peptide corresponding to the N-terminus of the Human NEK10. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction