missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEIL2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA584913
This item is not returnable.
View return policy
Description
Immunogen sequence: DPSPRLVLHF GGGGFLAFYN CQLSWSSSPV VTPTCDILSE KFHRGQALEA LGQAQPVCYT LLDQRYFSGL GN
Involved in base excision repair of DNA damaged by oxidation or by mutagenic agents. Has DNA glycosylase activity towards 5-hydroxyuracil and other oxidized derivatives of cytosine with a preference for mismatched double-stranded DNA (DNA bubbles). Has low or no DNA glycosylase activity towards thymine glycol, 2-hydroxyadenine, hypoxanthine and 8-oxoguanine. Has AP (apurinic/apyrimidinic) lyase activity and introduces nicks in the DNA strand. Cleaves the DNA backbone by beta-delta elimination to generate a single-strand break at the site of the removed base with both 3'- and 5'-phosphates.
Specifications
| NEIL2 | |
| Polyclonal | |
| Unconjugated | |
| NEIL2 | |
| DNA glycosylase/AP lyase Neil2; DNA-(apurinic or apyrimidinic site) lyase Neil2; Endonuclease 8-like 2; endonuclease VIII-like 2; Gm1212; NEH2; nei endonuclease VIII-like 2; nei endonuclease VIII-like 2 (E. coli); nei homolog 2; nei like 2; nei like 2 (E. coli); nei like DNA glycosylase 2; NEI2; Neil2; nei-like DNA glycosylase 2; nei-like protein 2 | |
| Rabbit | |
| Anitgen affinity chromatography, Antigen affinity chromatography | |
| RUO | |
| 252969 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| Lot-Specific | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q969S2 | |
| NEIL2 | |
| Recombinant protein corresponding to Human NEIL2 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction