Learn More
NEIL1 Polyclonal Antibody
GREENER_CHOICE
NEIL1 Polyclonal Antibody
Brand:  Invitrogen PA596438
Product Code. 16599271
  • £442.00 / 100µL
Description
NEIL1 Polyclonal Antibody for ICC/IF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NEIL1
Polyclonal
Unconjugated
NEIL1
2810450N13Rik; DNA endonuclease eight-like glycosylase 1; DNA glycosylase/AP lyase Neil1; DNA-(apurinic or apyrimidinic site) lyase Neil1; endonuclease 8-like 1; endonuclease VIII; endonuclease VIII-like 1; FLJ22402; FPG1; hFPG1; NEH1; nei endonuclease VIII-like 1; nei endonuclease VIII-like 1 (E. coli); nei homolog 1; nei like DNA glycosylase 1; Nei1; NEIL1; nei-like DNA glycosylase 1; nei-like protein 1
Rabbit
Affinity Chromatography
RUO
367090, 72774, 79661
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
0.58 mg/mL
PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Q8K4Q6, Q96FI4
NEIL1
Recombinant fusion protein containing a sequence corresponding to amino acids 1-135 of human NEIL1 (NP_0788842).
100 μL
Primary
Human, Mouse, Rat