Learn More
NEGR1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA552870
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: AGGDKWSVDP RVSISTLNKR DYSLQIQNVD VTDDGPYTCS VQTQHTPRTM QVHLTVQVPP KIYDISNDMT VNEGTNVTLT CLATGKPEPS ISWRHISPSA KPFENGQYLD IYGITRDQAG EYECSA Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
May be involved in cell-adhesion. May function as a trans-neural growth-promoting factor in regenerative axon sprouting in the mammalian brain.Specifications
NEGR1 | |
Polyclonal | |
Unconjugated | |
NEGR1 | |
5330422G01Rik; a kindred of IgLON; DMML2433; IgLON family member 4; IGLON4; Kiaa3001; KILON; kilon protein; kindred of IgLON; NEGR1; Neuronal growth regulator 1; neurotractin; Ntra; UNQ2433; UNQ2433/PRO4993 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
257194 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.09 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7Z3B1 | |
NEGR1 | |
Recombinant protein corresponding to Human NEGR1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only