missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NEGR1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5148440
This item is not returnable.
View return policy
Description
NEGR1 Polyclonal Antibody for Western Blot, IP, ELISA
May be involved in cell-adhesion. May function as a trans-neural growth-promoting factor in regenerative axon sprouting in the mammalian brain.
Specifications
| NEGR1 | |
| Polyclonal | |
| Unconjugated | |
| NEGR1 | |
| 5330422G01Rik; a kindred of IgLON; DMML2433; IgLON family member 4; IGLON4; Kiaa3001; KILON; kilon protein; kindred of IgLON; NEGR1; Neuronal growth regulator 1; neurotractin; Ntra; UNQ2433; UNQ2433/PRO4993 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 320840 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunoprecipitation, Western Blot | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q80Z24 | |
| NEGR1 | |
| Recombinant Mouse NEGR1 Protein (Met1-Gly318). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction