missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NEFL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA587394
This item is not returnable.
View return policy
Description
Immunogen sequence: SSGSLMPSLE NLDLSQVAAI SNDLKSIRTQ EKAQLQDLND RFASFIERVH ELEQQNKVLE AELLVLRQKH SEPSRFRALY EQEIRDLRLA AEDATNEKQA LQGEREGLEE TLRNLQARYE EEVLSREDAE GRLMEARKGA DEAALARAEL EKRIDSLMDE ISFLKKVHEE EIAELQAQIQ YAQISVEMDV T Positive Samples: MCF7.
Involved in the maintenance of neuronal caliber, neurofilaments are the intermediate filament proteins found specifically in neurons, and are composed predominantly of three major proteins called NF-L, NF-M and NF-H. Like most other intermediate filament proteins (IFPs), the expression of the different neuronal IFPs is both tissue-specific and developmentally regulated. NF-L is the light or low molecular weight microfilament subunit and runs on SDS-PAGE gels at approximately 70 kDa. Neurofilament are the 10nm or intermediate filament proteins found specifically in neurons, and are composed predominantly of three major proteins called NF-L, NF-M and NF-H. NF-H is the heavy or high molecular weight microfilament subunit and runs on SDS-PAGE gels in the range 180-220 kDa, with some variation in different species.
Specifications
| NEFL | |
| Polyclonal | |
| Unconjugated | |
| NEFL | |
| 68 kDa neurofilament protein; AI847934; CMT1F; CMT2E; FLJ53642; light molecular weight neurofilament protein; micro glutamic acid-rich protein; NEF3; NEFL; nefl.L; neurofilament; neurofilament 3; neurofilament L subunit; neurofilament light; neurofilament light polypeptide; neurofilament protein; neurofilament protein L; neurofilament protein, light chain; neurofilament subunit NF-L; neurofilament triplet L protein; neurofilament, light L homeolog; neurofilament, light polypeptide; neurofilament, light polypeptide 68kDa; neurofilament, light polypeptide L homeolog; neurofilament-l; neurofilament-light; Nf68; NFL; N-FL; NF-L; NFM; NF-M; NLC; OTTHUMP00000196614; PPP1R110; protein phosphatase 1, regulatory subunit 110; XELAEV_18018673mg; XNF-L | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 18039, 4747, 83613 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P07196, P08551, P19527 | |
| NEFL | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 60-250 of human Neurofilament L (NP_0061492). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction