missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEFL Monoclonal Antibody (6H112), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA547465
This item is not returnable.
View return policy
Description
Involved in the maintenance of neuronal caliber, neurofilaments are the intermediate filament proteins found specifically in neurons, and are composed predominantly of three major proteins called NF-L, NF-M and NF-H. Like most other intermediate filament proteins (IFPs), the expression of the different neuronal IFPs is both tissue-specific and developmentally regulated. NF-L is the light or low molecular weight microfilament subunit and runs on SDS-PAGE gels at approximately 70 kDa. Neurofilament are the 10nm or intermediate filament proteins found specifically in neurons, and are composed predominantly of three major proteins called NF-L, NF-M and NF-H. NF-H is the heavy or high molecular weight microfilament subunit and runs on SDS-PAGE gels in the range 180-220 kDa, with some variation in different species.Specifications
NEFL | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 5mM sodium azide | |
P02547, P02548, P07196, P08551, P19527 | |
NEFL | |
C-terminal peptide of human NF-L protein, GEEEDTKESEEEEKKEESAGEEQVAKKKD with an N-terminal C for coupling to KLH | |
100 μL | |
Primary | |
Bovine, Human, Mouse, Porcine, Rat | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunohistochemistry (PFA fixed), Western Blot | |
6H112 | |
Unconjugated | |
NEFL | |
68 kDa neurofilament protein; AI847934; CMT1F; CMT2E; FLJ53642; light molecular weight neurofilament protein; micro glutamic acid-rich protein; NEF3; Nefl; nefl.L; neurofilament; neurofilament 3; neurofilament L subunit; neurofilament light; neurofilament light polypeptide; neurofilament protein; neurofilament protein L; neurofilament protein, light chain; neurofilament subunit NF-L; neurofilament triplet L protein; neurofilament, light L homeolog; neurofilament, light polypeptide; neurofilament, light polypeptide 68kDa; neurofilament, light polypeptide L homeolog; neurofilament-l; neurofilament-light; Nf68; NFL; NF-L; NFM; NF-M; NLC; OTTHUMP00000196614; PPP1R110; protein phosphatase 1, regulatory subunit 110; XELAEV_18018673mg; XNF-L | |
Mouse | |
Protein A/G | |
RUO | |
100521224, 18039, 281348, 4747, 83613 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |