missing translation for 'onlineSavingsMsg'
Learn More

Nefiracetam, MedChemExpress

Product Code. p-200096162
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
This item is not returnable. View return policy

Product Code. 16468246

Brand: Medchem Express HYB0340

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Nefiracetam is a GABAergic, cholinergic, and monoaminergic neuronal systems enhancer for Ro 5-4864-induced convulsions.

Nefiracetam is a GABAergic, cholinergic, and monoaminergic neuronal systems enhancer for Ro 5-4864-induced convulsions.

Specifications

Chemical Name or Material Nefiracetam
CAS 77191-36-7
Color White
Purity Grade Notes Research
Molecular Formula C14H18N2O2
Quantity 100 mg
Synonym DM9384 DZL-221
Solubility Information DMSO : ≥ 100 mg/mL (406.01 mM)
SMILES O=C(NC1=C(C)C=CC=C1C)CN2C(CCC2)=O
Molecular Weight (g/mol) 246.3
Formula Weight 246.3
Percent Purity 99.39%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.