missing translation for 'onlineSavingsMsg'
Learn More

Nefazodone hydrochloride, MedChemExpress

Product Code. p-200099609
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16478698

Brand: Medchem Express HYB1396

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Nefazodone hydrochloride (BMY-13754) is a potent and selective 5HT2A (Ki=5.8 nM) antagonist with moderate inhibition of 5-HT and noradrenaline uptake (IC50 of 290 and 300 nM, respectively). Nefazodone hydrochloride is a phenylpiperazine antidepressant with less alpha-adrenergic blocking activity.

Nefazodone hydrochloride (BMY-13754) is a potent and selective 5HT2A (Ki=5.8 nM) antagonist with moderate inhibition of 5-HT and noradrenaline uptake (IC50 of 290 and 300 nM, respectively). Nefazodone hydrochloride is a phenylpiperazine antidepressant with less alpha-adrenergic blocking activity.

Specifications

Chemical Name or Material Nefazodone hydrochloride
CAS 82752-99-6
Color White
Purity Grade Notes Research
Molecular Formula C25H33Cl2N5O2
Quantity 10 mg
Synonym BMY-13754 MJ-13754-1
Solubility Information DMSO : 50 mg/mL (98.72 mM; Need ultrasonic) ∣H2O : 2 mg/mL (3.95 mM; Need ultrasonic)
SMILES O=C1N(CCCN2CCN(C3=CC=CC(Cl)=C3)CC2)N=C(CC)N1CCOC4=CC=CC=C4.[H]Cl
Molecular Weight (g/mol) 506.47
Formula Weight 506.47
Percent Purity 99.02%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.