Learn More
Needle Cleaning Solution
Product Code. 11930557
Change view
Click to view available options
For Use With (Equipment):
Osmometer Type 15/15M and 16/16M
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
11930557 Osmometer Type 15/15M and 16/16M Each
1 options
Product Code. 11930557 Supplier Loeser Supplier No. 0.01.0004
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Needle cleaning solution for i Osmometer
  • Solution for cleaning the needle hole of automatic Osmometers
  • 20mL bottle
Specifications
Product Type Needle Cleaning Solution
For Use With (Equipment) Osmometer Type 15/15M and 16/16M
Volume (Metric) 20 mL