missing translation for 'onlineSavingsMsg'
Learn More

NEDD4L Rabbit anti-Human, Clone: 3W9J2, Novus Biologicals™

Product Code. 18377852 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18377852 100 μg 100µL
18304722 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18377852 Supplier Novus Biologicals Supplier No. NBP316747100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

NEDD4L Monoclonal antibody specifically detects NEDD4L in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen NEDD4L
Applications Western Blot
Classification Monoclonal
Clone 3W9J2
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias E3 ubiquitin-protein ligase NEDD4-like, EC 6.3.2, EC 6.3.2.-, FLJ33870, hNedd4-2, KIAA0439ubiquitin-protein ligase NEDD4-like, NEDD4.2, Nedd4-2, NEDL3, neural precursor cell expressed, developmentally down-regulated 4-like, neural precursor cell expressed, developmentally down-regulated gene 4-like, RSP5ubiquitin-protein ligase Rsp5
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 876-975 of human NEDD4L (Q96PU5).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology, Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 23327
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.