missing translation for 'onlineSavingsMsg'
Learn More

NEDD4L Antibody (1D2), Novus Biologicals™

Product Code. 18376558 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18376558 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18376558 Supplier Novus Biologicals Supplier No. H00023327M04

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NEDD4L Monoclonal antibody specifically detects NEDD4L in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen NEDD4L
Applications Western Blot, ELISA, Immunoprecipitation, Sandwich ELISA
Classification Monoclonal
Clone 1D2
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunoprecipitation, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH32597
Gene Alias E3 ubiquitin-protein ligase NEDD4-like, EC 6.3.2, EC 6.3.2.-, FLJ33870, hNedd4-2, KIAA0439ubiquitin-protein ligase NEDD4-like, NEDD4.2, Nedd4-2, NEDL3, neural precursor cell expressed, developmentally down-regulated 4-like, neural precursor cell expressed, developmentally down-regulated gene 4-like, RSP5ubiquitin-protein ligase Rsp5
Host Species Mouse
Immunogen NEDD4L (AAH32597, 1 a.a. ~ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MATGLGEPVYGLSEDEGESRILRVKVVSGIDLAKKDIFGASDPYVKLSLYVADENRELALVQTKTIKKTLNPKWNEEFYFRVNPSNHRLLFEVFDENRLT
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology, Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 23327
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.