missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEDD4 Recombinant Rabbit Monoclonal Antibody (23GB2255), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen MA552487
This item is not returnable.
View return policy
Description
NEDD4 Recombinant Monoclonal antibody specifically detects NEDD4 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot
Specifications
| NEDD4 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| NEDD4 | |
| AA959633; AL023035; AU019897; cell proliferation-inducing gene 53 protein; E3 ubiquitin-protein ligase NEDD4; E430025J12Rik; HECT-type E3 ubiquitin transferase NEDD4; Kiaa0093; mKIAA0093; NEDD4; NEDD-4; NEDD4 E3 ubiquitin protein ligase; Nedd4-1; Nedd4a; Neural precursor cell expressed developmentally down-regulated protein 4; neural precursor cell expressed, developmentally down-regulated 4; neural precursor cell expressed, developmentally down-regulated 4, E3 ubiquitin protein ligase; neural precursor cell expressed, developmentally down-regulated gene 4; neural precursor cell expressed, developmentally down-regulated gene 4a; neural precursor cell expressed, developmentally down-regulted gene 4; OTTHUMP00000217840; OTTHUMP00000217841; OTTHUMP00000217842; PIG53; receptor-potentiating factor 1; RPF1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 17999, 25489, 4734 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB2255 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P46934, P46935, Q62940 | |
| NEDD4 | |
| A synthesized peptide derived from human NEDD4 (1271-1319AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction