missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEDD4 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97422PCP
This item is not returnable.
View return policy
Description
NEDD4 Polyclonal antibody specifically detects NEDD4 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| NEDD4 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| Cell proliferation-inducing gene 53 protein, E3 ubiquitin-protein ligase NEDD4, EC 6.3.2, EC 6.3.2.-, KIAA0093NEDD4-1MGC176705, NEDD-4, Neural precursor cell expressed developmentally down-regulated protein 4, neural precursor cell expressed, developmentally down-regulated 4, receptor-potentiating factor 1, RPF1 | |
| Produced in rabbits immunized with E. coli-derived Human NEDD4 fragment. | |
| 0.1 mL | |
| Cell Biology, Cell Cycle and Replication, Endocrinology, Immunology, Neuroscience, Signal Transduction | |
| 4734 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| PerCP | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction