missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
NEDD4 Polyclonal antibody specifically detects NEDD4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | NEDD4 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Formulation | PBS (pH 7.2) and 40% Glycerol |
| Gene Alias | Cell proliferation-inducing gene 53 protein, E3 ubiquitin-protein ligase NEDD4, EC 6.3.2, EC 6.3.2.-, KIAA0093NEDD4-1MGC176705, NEDD-4, Neural precursor cell expressed developmentally down-regulated protein 4, neural precursor cell expressed, developmentally down-regulated 4, receptor-potentiating factor 1, RPF1 |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids: TVCEKSSEGYSCVSVHFTQRKAATLDCETTNGDCKPEMSEIKLHSDSEYIKLMHRTSACLPSSQNVDCQININGELERPHSQMNKNHGILRRSISLG |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?