missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEDD4 Antibody (JE35-58), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32641
This item is not returnable.
View return policy
Description
NEDD4 Monoclonal antibody specifically detects NEDD4 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| NEDD4 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000 | |
| Cell proliferation-inducing gene 53 protein, E3 ubiquitin-protein ligase NEDD4, EC 6.3.2, EC 6.3.2.-, KIAA0093NEDD4-1MGC176705, NEDD-4, Neural precursor cell expressed developmentally down-regulated protein 4, neural precursor cell expressed, developmentally down-regulated 4, receptor-potentiating factor 1, RPF1 | |
| Recombinant fragment corresponding to Human NEDD4 aa 160-460. (Uniprot: P46934) | |
| 100 μL | |
| Cell Biology, Cell Cycle and Replication, Endocrinology, Immunology, Neuroscience, Signal Transduction | |
| 4734 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| JE35-58 | |
| Unconjugated | |
| 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction