missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ NED™ Matrix Standard

Product Code. 16325262 Shop All Applied Biosystems Products
Change view
Click to view available options
For Use With (Equipment):
310 Genetic Analyzer
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
16325262 310 Genetic Analyzer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16325262 Supplier Applied Biosystems™ Supplier No. 402996

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

1 Tube of NED Matrix Standard

Uses the multicomponent matrix to automatically analyze the four different colored fluorescent dye-labeled samples in a single capillary.

The NED™ Matrix Standard can be used to analyze 6-FAM™, VIC®, NED™ and ROX™-labeled DNA fragments in fragment analysis applications. The standard can be used on the Applied Biosystems® 310 Genetic Analyzers.

Order Info

Each

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains 1 tube of NED Matrix Standard. Store at 2-8°C. (Do not freeze).
For Use With (Equipment) 310 Genetic Analyzer
For Use With (Application) Sequencing
Label or Dye NED
Product Line NED
Product Type Matrix Standard
Quantity 1 Tube
Shipping Condition Wet Ice

For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.