missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Nectin-4/PVRL4 Antibody (enfortumab), Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28108-100ug
This item is not returnable.
View return policy
Description
Nectin-4/PVRL4 Monoclonal antibody specifically detects Nectin-4/PVRL4 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Nectin-4/PVRL4 | |
| Monoclonal | |
| Unconjugated | |
| Q96NY8 | |
| Human | |
| Protein A purified | |
| RUO | |
| 81607 | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| enfortumab | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| EDSS1, Ig superfamily receptor LNIR, LNIRnectin 4, nectin-4, poliovirus receptor-related 4, poliovirus receptor-related protein 4, PRR4DKFZp686K05193 | |
| Nectin-4 | |
| 100 μg | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction