missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nectin-4/PVRL4 Antibody (337516), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB2659H
407.15 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
Nectin-4/PVRL4 Monoclonal antibody specifically detects Nectin-4/PVRL4 in Human samples. It is validated for Flow Cytometry, CyTOF
Tekniske data
| Nectin-4/PVRL4 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 81607 | |
| Store at 4C in the dark. | |
| IgG2b |
| Flow Cytometry, CyTOF | |
| 337516 | |
| Flow Cytometry, CyTOF-ready | |
| EDSS1, Ig superfamily receptor LNIR, LNIRnectin 4, nectin-4, poliovirus receptor-related 4, poliovirus receptor-related protein 4, PRR4DKFZp686K05193 | |
| Mouse myeloma cell line NS0-derived recombinant human Nectin-4, Gly27-Val351, Accession # Q96NY8 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion