missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Nectin 2 Recombinant Rabbit Monoclonal Antibody (005), PE

Description
Nectin 2 Recombinant Monoclonal Antibody for Flow
This gene encodes a single-pass type I membrane glycoprotein with two Ig-like C2-type domains and an Ig-like V-type domain. This protein is one of the plasma membrane components of adherens junctions. It also serves as an entry for certain mutant strains of herpes simplex virus and pseudorabies virus, and it is involved in cell to cell spreading of these viruses. Variations in this gene have been associated with differences in the severity of multiple sclerosis. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications
Specifications
| Antigen | Nectin 2 |
| Applications | Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Clone | 005 |
| Concentration | 0.1 mg/mL |
| Conjugate | PE |
| Formulation | PBS with 0.5% BSA and 0.03% ProClin 300 |
| Gene | NECTIN2 |
| Gene Accession No. | Q92692 |
| Gene Alias | AI325026; AI987993; CD112; herpes virus entry mediator B; Herpesvirus entry mediator B; herpesvirus entry protein B; hveB; mHveB; MPH; Murine herpes virus entry protein B; murine herpesvirus entry protein B; nectin cell adhesion molecule 2; Nectin2; Nectin-2; Poliovirus receptor homolog; poliovirus receptor related 2; poliovirus receptor-like 2; poliovirus receptor-related 2; poliovirus receptor-related 2 (herpesvirus entry mediator B); poliovirus receptor-related protein 2; poliovirus sensitivity; PRR2; Pvr; Pvrl2; PVRR2; Pvs |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?