missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PAR1 Monoclonal Antibody (1028505)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561350
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Thrombin is a serine protease that is involved in platelet aggregation and blood coagulation. It is cleaved from its precursor, prothrombin, and converts fibrinogen to fibrin in the final step of the clotting cascade. Thrombin mediates its regulatory effects by activating cell surface G-protein coupled receptors which include PAR-1, PAR-2 and PAR3.
Specifications
| PAR1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P25116 | |
| F2R | |
| Human PAR1 synthetic peptide. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin) | |
| 1028505 | |
| Unconjugated | |
| F2R | |
| AI482343; C HTR; Cf2r; coagulation factor II (thrombin) receptor; coagulation factor II receptor; coagulation factor II thrombin receptor; F2R; HTR; PAR1; PAR-1; protease activated receptor 1; protease-activated receptor 1; proteinase-activated receptor 1; Thrombin receptor; ThrR; TR; TRGPC | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 2149 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering