missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Nectin 2 Monoclonal Antibody (R2.525)
GREENER_CHOICE

Product Code. 11525092
Change view
Click to view available options
Quantity:
200 μg
Unit Size:
200µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11525092 200 μg 200µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11525092 Supplier Invitrogen™ Supplier No. MA135890

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Mouse anti Human CD112 antibody, clone R2-525 recognizes CD112, also known as Poliovirus Receptor Related 2, PRR2 or Nectin-2, a surface molecule originally reported to be homologous to CD155, the poliovirus receptor (Lopez et al. 1998). PRR2 expression is restricted to the myelo-monocytic and megakaryocytic lineages. PRR2 expression has also been detected on haematopoeitic progenitors expressing CD34.

This gene encodes a single-pass type I membrane glycoprotein with two Ig-like C2-type domains and an Ig-like V-type domain. This protein is one of the plasma membrane components of adherens junctions. It also serves as an entry for certain mutant strains of herpes simplex virus and pseudorabies virus, and it is involved in cell to cell spreading of these viruses. Variations in this gene have been associated with differences in the severity of multiple sclerosis. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nectin 2
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone R2.525
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene NECTIN2
Gene Accession No. Q92692
Gene Alias AI325026; AI987993; CD112; herpes virus entry mediator B; Herpesvirus entry mediator B; herpesvirus entry protein B; hveB; mHveB; MPH; Murine herpes virus entry protein B; murine herpesvirus entry protein B; nectin cell adhesion molecule 2; Nectin2; Nectin-2; Poliovirus receptor homolog; poliovirus receptor related 2; poliovirus receptor-like 2; poliovirus receptor-related 2; poliovirus receptor-related 2 (herpesvirus entry mediator B); poliovirus receptor-related protein 2; poliovirus sensitivity; PRR2; Pvr; Pvrl2; PVRR2; Pvs
Gene Symbols NECTIN2
Host Species Mouse
Immunogen Proprietary immunogen.
Purification Method purified
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5819
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.