missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nectin-2/CD112 Rabbit anti-Human, Clone: 5, Novus Biologicals™
Rabbit Monoclonal Antibody
£296.00 - £429.00
Specifications
| Antigen | Nectin-2/CD112 |
|---|---|
| Clone | 5 |
| Dilution | Flow Cytometry 1:25-1:100, Sandwich ELISA Capture 1:250-1:2000 |
| Applications | Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18249147
|
Novus Biologicals
NBP2-89188-50UL |
50 μg |
£296.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18264846
|
Novus Biologicals
NBP2-89188-100UL |
100 μg |
£429.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nectin-2/CD112 Monoclonal specifically detects Nectin-2/CD112 in Human samples. It is validated for Flow Cytometry, Sandwich ELISA Capture.Specifications
| Nectin-2/CD112 | |
| Flow Cytometry 1:25-1:100, Sandwich ELISA Capture 1:250-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 5819 | |
| IgG | |
| Protein A purified |
| 5 | |
| Flow Cytometry, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| CD112, CD112 antigen, Herpes virus entry mediator B, Herpesvirus entry mediator B, herpesvirus entry protein B, HveB, HVEBpoliovirus receptor-like 2, nectin 2, Nectin-2, poliovirus receptor-related 2 (herpesvirus entry mediator B), poliovirus receptor-related protein 2, PRR2nectin-2, PVRR2poliovirus receptor related 2 | |
| Recombinant Human Nectin-2/CD112 protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title