missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nectin-2/CD112 Antibody (R2.525) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-09163
This item is not returnable.
View return policy
Description
Nectin-2/CD112 Monoclonal specifically detects Nectin-2/CD112 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation, Immunohistochemistry-Frozen.
Specifications
| Nectin-2/CD112 | |
| Monoclonal | |
| 1 mg/ml | |
| Flow Cytometry 1-4 ug/ml, Immunoprecipitation, Immunohistochemistry-Frozen | |
| CD112, CD112 antigen, Herpes virus entry mediator B, Herpesvirus entry mediator B, herpesvirus entry protein B, HveB, HVEBpoliovirus receptor-like 2, nectin 2, Nectin-2, poliovirus receptor-related 2 (herpesvirus entry mediator B), poliovirus receptor-related protein 2, PRR2nectin-2, PVRR2poliovirus receptor related 2 | |
| NIH/3T3 cells transfected with human Nectin-2 | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Frozen) | |
| R2.525 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 5819 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction