missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This solid phase sandwich ELISA utilizes an antibody specific for Human Nectin-2/CD112.
Specifications
Specifications
| Assay Range | 15.625 pg/mL |
| Assay Sensitivity | 15.625-1000 pg/mL |
| Assay | Sandwich ELISA |
| Conjugate | HRP |
| Product Type | Antibody Pairs |
| Specificity | This solid phase sandwich ELISA utilizes an antibody specific for Human Nectin-2/CD112. |
| Target Species | Human |
| For Use With (Application) | Sandwich ELISA |
| Gene Alias | CD112, CD112 antigen, Herpes virus entry mediator B, Herpesvirus entry mediator B, herpesvirus entry protein B, HveB, HVEBpoliovirus receptor-like 2, nectin 2, Nectin-2, poliovirus receptor-related 2 (herpesvirus entry mediator B), poliovirus receptor-related protein 2, PRR2nectin-2, PVRR2poliovirus receptor related 2 |
| Gene ID (Entrez) | 5819 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?