missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nectin-1/PVRL1 Antibody (OTI6F8) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72929
This item is not returnable.
View return policy
Description
Nectin-1/PVRL1 Monoclonal specifically detects Nectin-1/PVRL1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Nectin-1/PVRL1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:2000, Immunohistochemistry 1:150, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:150 | |
| CD111, CD111 antigen, CLPED1ectodermal dysplasia 4 (Margarita Island type), ED4, Herpes virus entry mediator C, Herpesvirus entry mediator C, Herpesvirus Ig-like receptor, HIgRPVRR, HveC, HVECpoliovirus receptor-like 1, MGC142031, Nectin-1, OFC7nectin 1, poliovirus receptor-related 1 (herpesvirus entry mediator C), poliovirus receptor-related protein 1, PRR, PRR1MGC16207, PVRR1nectin-1, SK-12 | |
| Mouse | |
| 54 kDa | |
| 100 μg | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| OTI6F8 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| NECTIN1 | |
| Full length human recombinant protein of human PVRL1(NP_002846) produced in HEK293T cell. | |
| Affinity Purified | |
| Neuroscience | |
| 5818 | |
| Human | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction