missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nectin-1 (CD111) Mouse anti-Human, BUV661, Clone: CK41, BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 750764
This item is not returnable.
View return policy
Description
Nectin-1 (CD111) Monoclonal antibody specifically detects Nectin-1 (CD111) in Human samples. It is validated for Flow Cytometry
Specifications
| Nectin-1 (CD111) | |
| Monoclonal | |
| Brilliant Ultraviolet 661 | |
| Nectin1; PRR1; PVRL1; PVRR1; HveC; HIgR; CLPED1; ED4; SK-12; OFC7 | |
| Human Recombinant Protein | |
| RUO | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| CK41 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV661 under optimal conditions that minimize unconjugated dye and antibody. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction