missing translation for 'onlineSavingsMsg'
Learn More

NecroX-5, MedChemExpress

Product Code. 16433928
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16433928 10 mg 10mg
16430287 50 mg 50mg
16426647 5 mg 5mg
16422916 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16433928 Supplier Medchem Express Supplier No. HY104015
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress NecroX-5 is a derivative of the NecroX, reduces intracellular calcium concentration, and possesses anti-inflammatory and anti-cancer activity.

NecroX-5 is a derivative of the NecroX, reduces intracellular calcium concentration, and possesses anti-inflammatory and anti-cancer activity.

Specifications

Chemical Name or Material NecroX-5
CAS 1383718-29-3
Color Light Brown to Black
Purity Grade Notes Research
Molecular Formula C27H39N3O9S3
Quantity 10 mg
Solubility Information DMSO : 170 mg/mL (263.24 mM; Need ultrasonic)
SMILES O=S1(CCN(CC2=CC3=C(NC(C4=CC=CC=C4)=C3)C(NCC5CCOCC5)=C2)CC1)=O.CS(=O)(O)=O.CS(=O)(O)=O
Molecular Weight (g/mol) 645.81
Formula Weight 645.81
Percent Purity 98.52%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.