missing translation for 'onlineSavingsMsg'
Learn More

Necrostatin 2 S enantiomer, MedChemExpress

Product Code. 16456118
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
5mg
Denna artikel kan inte returneras. Se returpolicy

Produktkod. 16456118

Brand: Medchem Express HY14622B

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

MedChemExpress Necrostatin 2 S enantiomer is the S enantiomer of Necrostatin 2.

Necrostatin 2 S enantiomer is the S enantiomer of Necrostatin 2.

Specifikationer

Chemical Name or Material Necrostatin 2 S enantiomer
Physical Form Solid
CAS 852391-20-9
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C13H12ClN3O2
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 50 mg/mL (180.04 mM; Need ultrasonic)
SMILES O=C1N(C)C([C@H](CC2=CNC3=C2C=CC=C3Cl)N1)=O
Molecular Weight (g/mol) 277.71
Formula Weight 277.71
Percent Purity 99.58%
Grade Research
Visa mer Visa mindre

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.