missing translation for 'onlineSavingsMsg'
Learn More

Necrostatin-1, MedChemExpress

Product Code. p-200099503
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16496916

Brand: Medchem Express HY15760

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Necrostatin-1 (Nec-1) is a potent and cross the blood-brain barrier necroptosis inhibitor with an EC50 of 490 nM in Jurkat cells. Necrostatin-1 inhibits RIP1 kinase (EC50=182 nM). Necrostatin-1 is also an IDO inhibitor.

Necrostatin-1 (Nec-1) is a potent and cross the blood-brain barrier necroptosis inhibitor with an EC50 of 490 nM in Jurkat cells. Necrostatin-1 inhibits RIP1 kinase (EC50=182 nM). Necrostatin-1 is also an IDO inhibitor.

Specifications

Chemical Name or Material Necrostatin-1
Physical Form Powder
CAS 4311-88-0
Color Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C13H13N3OS
For Use With (Application) Cancer-Kinase/protease
Synonym Nec-1
Solubility Information DMSO : ≥ 46 mg/mL (177.38 mM)
SMILES O=C(C(CC1=CNC2=C1C=CC=C2)N3)N(C)C3=S
Molecular Weight (g/mol) 259.33
Formula Weight 259.33
Percent Purity 98.96%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.