missing translation for 'onlineSavingsMsg'
Learn More

Necrostatin-1, MedChemExpress

Product Code. 16496916
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16496916 100 mg 100mg
16422877 50 mg 50mg
16406808 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16496916 Supplier Medchem Express Supplier No. HY15760
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Necrostatin-1 (Nec-1) is a potent and cross the blood-brain barrier necroptosis inhibitor with an EC50 of 490 nM in Jurkat cells. Necrostatin-1 inhibits RIP1 kinase (EC50=182 nM). Necrostatin-1 is also an IDO inhibitor.

Necrostatin-1 (Nec-1) is a potent and cross the blood-brain barrier necroptosis inhibitor with an EC50 of 490 nM in Jurkat cells. Necrostatin-1 inhibits RIP1 kinase (EC50=182 nM). Necrostatin-1 is also an IDO inhibitor.

Specifications

Chemical Name or Material Necrostatin-1
Physical Form Powder
CAS 4311-88-0
Color Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C13H13N3OS
For Use With (Application) Cancer-Kinase/protease
Synonym Nec-1
Solubility Information DMSO : ≥ 46 mg/mL (177.38 mM)
SMILES O=C(C(CC1=CNC2=C1C=CC=C2)N3)N(C)C3=S
Molecular Weight (g/mol) 259.33
Formula Weight 259.33
Percent Purity 98.96%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.