missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Necitumumab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA559394
This item is not returnable.
View return policy
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Necitumumab is an intravenously administered recombinant monoclonal IgG1 antibody used in the treatment of non-small cell lung cancer (NSCLC) as an EGFR antagonist. It functions by binding to epidermal growth factor receptor (EGFR) and prevents binding of its ligands, a process that is involved in cell proliferation, metastasis, angiogenesis, and malignant progression. Binding of necitumumab to EGFR induces receptor internalization and degradation, thereby preventing further activation of EGFR which is beneficial in NSCLC as many patients have increased protein expression of EGFR. Necitumumab is approved for use in combination with cisplatin and gemcitabine as a first-line treatment for metastatic squamous non-small cell lung cancer (NSCLC).
Specifications
| Necitumumab | |
| Recombinant Monoclonal | |
| Unconjugated | |
| 11F8; IMC-11F8; LY3012211 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Human | |
| 1 mg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction