missing translation for 'onlineSavingsMsg'
Learn More

Necdin Antibody (OTI5D1), DyLight 594, Novus Biologicals™

Product Code. 18422864 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18422864 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18422864 Supplier Novus Biologicals Supplier No. NBP272925DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Necdin Monoclonal antibody specifically detects Necdin in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Necdin
Applications Western Blot, Immunofluorescence, Immunofluorescence
Classification Monoclonal
Clone OTI5D1
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias HsT16328, necdin, necdin (mouse) homolog, necdin homolog (mouse), PWCR
Host Species Mouse
Immunogen Full length recombinant protein of human Necdin (NP_002478) produced in HEK293T cells.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 4692
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.