missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NECAP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15899-1-AP-20UL
This item is not returnable.
View return policy
Specifications
| NECAP2 | |
| Polyclonal | |
| Unconjugated | |
| NECAP2 | |
| NECAP 2, NECAP endocytosis associated 2, NECAP2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 298598, 55707, 66147 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6P756, Q9D1J1, Q9NVZ3 | |
| NECAP2 | |
| NECAP2 Fusion Protein Ag8721 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction