missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NECAP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £484.00
Specifications
| Antigen | NECAP1 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18232732
|
Novus Biologicals
NBP2-55494 |
100 μL |
£484.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18608357
|
Novus Biologicals
NBP2-55494-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NECAP1 Polyclonal specifically detects NECAP1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| NECAP1 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 25977 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:GQTIKLCIGNITNKKGGASKPRTA | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| adaptin ear-binding coat-associated protein 1, adaptin-ear-binding coat-associated protein 1, DKFZP566B183, MGC131900, NECAP endocytosis associated 1, NECAP endocytosis-associated protein 1, NECAP-1 | |
| NECAP1 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title