missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NECAB3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18940-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene interacts with the amino-terminal domain of the neuron-specific X11-like protein (X11L), inhibits the association of X11L with amyloid precursor protein through a non-competitive mechanism, and abolishes the suppression of beta-amyloid production by X11L. This protein, together with X11L, may play an important role in the regulatory system of amyloid precursor protein metabolism and beta-amyloid generation. The protein is phosphorylated by NIMA-related expressed kinase 2, and localizes to the Golgi apparatus. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
| NECAB3 | |
| Polyclonal | |
| Unconjugated | |
| NECAB3 | |
| APBA2BP, EFCBP3, NECAB3, Nek2 interacting protein 1, NIP1, STIP3, SYTIP2, X11L binding protein 51, XB51 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 311562, 56846, 63941 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96P71, Q9D6J4 | |
| Necab3 | |
| NECAB3 Fusion Protein Ag13493 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction