missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NECAB3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-56551
This item is not returnable.
View return policy
Description
NECAB3 Polyclonal specifically detects NECAB3 in Human samples. It is validated for Western Blot.
Specifications
| NECAB3 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| NECAB3 | |
| Synthetic peptides corresponding to NECAB3 (N-terminal EF-hand calcium binding protein 3) The peptide sequence was selected from the N terminal of NECAB3)(50ug). Peptide sequence MACAGLLTVCLLRPPAPQPQPQTPRHPQLAPDPGPAGHTLFQDVFRRADK. | |
| 100 μL | |
| Lipid and Metabolism | |
| 63941 | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Yeast | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Amyloid beta A4 protein-binding family A member 2-binding protein, APBA2BPfamily A, member 2 binding protein, dJ63M2.4, dJ63M2.5, EFCBP3STIP3, EF-hand calcium binding protein 3, Neuronal calcium-binding protein 3, neuronal calcium-binding protein NECAB3, NIP1Nek2-interacting protein 1, N-terminal EF-hand calcium binding protein 3, N-terminal EF-hand calcium-binding protein 3, synaptotagmin interacting protein 2, synaptotagmin interacting protein STIP3, SYTIP2, XB51X11L-binding protein 51 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction