missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NECAB2 Recombinant Mouse Monoclonal Antibody (K8U020_8H5)

Product Code. 30827827
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30827827 100 μL
30825844 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30827827 Supplier Invitrogen™ Supplier No. MA564497

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Recombinant Monoclonal Antibody

Immunogen sequence: GTAVILDIFR RADKNDDGKL SLEEFQLFFA DGVLNEKELE DLFHTIDSDN TNHVDTKELC DYFVDHMGDY EDVLASLETL NHSVLKAMGY TKKVYEGGSN VDQFVTRFLL KETANQIQSL LSSVESAVEA IEEQTSQLRQ Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%, Pig - 96%, Cynomolgus monkey - 99%.

The protein encoded by this gene is a neuronal calcium-binding protein that binds to and modulates the function of at least two receptors, adenosine A(2A) receptor and metabotropic glutamate receptor type 5.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NECAB2
Applications Immunoprecipitation
Classification Recombinant Monoclonal
Clone K8U020_8H5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.09% sodium azide
Gene NECAB2
Gene Accession No. F1LQY6, Q7Z6G3
Gene Alias EF hand calcium binding protein 2; Efcbp2; EF-hand calcium binding protein 2; EF-hand calcium-binding protein 2; NECAB2; neuronal calcium binding 2; neuronal calcium-binding protein 2; N-terminal EF-hand calcium binding protein 2; N-terminal EF-hand calcium-binding protein 2; Stip-2; synaptotagmin-interacting protein 2
Gene Symbols NECAB2
Host Species Mouse
Immunogen Recombinant human NECAB2 (aa 61-200).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170928, 54550
Target Species Human, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.