Learn More
NECAB1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA555278
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: ASSQSGCLRI SIQKLSNESR YMIYEFWENS SVWNSHLQTN YSKTFQRSNV DFLETPELTS TMLVPASWW Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
NECAB1 is a protein coding gene.Specifications
NECAB1 | |
Polyclonal | |
Unconjugated | |
NECAB1 | |
1700003H21Rik; EF hand calcium binding protein 1; EFCBP1; EF-hand calcium binding protein 1; EF-hand calcium-binding protein 1; Necab1; neuronal Ca(2+)-binding protein 1; neuronal calcium binding protein; neuronal calcium-binding protein 1; N-terminal EF-hand calcium binding protein 1; N-terminal EF-hand calcium-binding protein 1; Stip; Stip-1; synaptotagmin interacting protein 1; synaptotagmin-interacting protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64168 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N987 | |
NECAB1 | |
Recombinant protein corresponding to Human NECAB1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only