Learn More
Invitrogen™ NECAB1 Monoclonal Antibody (CL0580)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524613
Description
Immunogen sequence: LLKETLNQLQ SLQNSLECAM ETTEEQTRQE RQGPAKPEVL SIQWPGKRSS RRVQRHNSFS PNSP Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%. Binds to an epitope located within the peptide sequence SIQWPGKRSS as determined by overlapping synthetic peptides.
NECAB1 is a protein coding gene.
Specifications
| NECAB1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N987 | |
| NECAB1 | |
| Recombinant protein fragment of Human NECAB1 (Product #RP-93776) | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| CL0580 | |
| Unconjugated | |
| NECAB1 | |
| 1700003H21Rik; EF hand calcium binding protein 1; EFCBP1; EF-hand calcium binding protein 1; EF-hand calcium-binding protein 1; Necab1; neuronal Ca(2+)-binding protein 1; neuronal calcium binding protein; neuronal calcium-binding protein 1; N-terminal EF-hand calcium binding protein 1; N-terminal EF-hand calcium-binding protein 1; Stip; Stip-1; synaptotagmin interacting protein 1; synaptotagmin-interacting protein 1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 64168 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.