Learn More
Thermo Scientific™ Nebulizers for ICP-OES Analyzers
Enhance your analysis capabilities with a range of nebulizers compatible with the Thermo Scientific™ ICP-OES analyzers.
Specifications
| For Use With (Equipment) | iCAP Series ICP-OES Systems, ICP-OES Analyzers |
|---|---|
| Type | Nebulizer |
Description
Enhance your analysis capabilities with a range of nebulizers compatible with the Thermo Scientific™ iCAP™ Series ICP-OES systems. Choose one to suit your sample matrix to ensure optimum performance.
Enhanced Performance through Advanced AccessoriesThese Nebulizers are available as standalone items, or as part of the Thermo Scientific™ ICP-OES Sample Introduction Kits.
- Standard Nebulizer handles nonaggressive liquid samples
- High Solids Nebulizer is intended for the analysis of samples containing relatively high solids (up to 15%)
- Burgener MiraMist Nebulizer, PEEK components handle most liquids, except for a few acids and organic solvents
- V Groove Nebulizer is intended for the analysis of samples containing very high solids (30% or more) and for analysis of nonvolatile organic solvents
Specifications
| iCAP Series ICP-OES Systems, ICP-OES Analyzers | |
| Nebulizer |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.