missing translation for 'onlineSavingsMsg'
Learn More

Nebivolol hydrochloride, MedChemExpress

Product Code. p-200101767
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16494297

Brand: Medchem Express HYB0203A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Nebivolol hydrochloride selectively inhibits β1- adrenergic receptor with IC50 of 0.8 nM.

Nebivolol hydrochloride selectively inhibits β1- adrenergic receptor with IC50 of 0.8 nM.

Specifications

Chemical Name or Material Nebivolol hydrochloride
CAS 152520-56-4
Color White
Purity Grade Notes Research
Molecular Formula C22H26ClF2NO4
Quantity 50 mg
Synonym R 065824 hydrochloride
Solubility Information DMSO : 100 mg/mL (226.30 mM; Need ultrasonic) ∣H2O : 1.79 mg/mL (4.05 mM; ultrasonic and warming and heat to 60°C)
SMILES FC1=CC=C2C(CC[C@]([C@@H](O)CNC[C@H](O)[C@@]3([H])CCC(C=C(F)C=C4)=C4O3)([H])O2)=C1.Cl
Molecular Weight (g/mol) 441.9
Formula Weight 441.9
Percent Purity 98.73%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.