missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nebivolol hydrochloride, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Nebivolol hydrochloride selectively inhibits β1- adrenergic receptor with IC50 of 0.8 nM.
Specifications
Specifications
| Chemical Name or Material | Nebivolol hydrochloride |
| CAS | 152520-56-4 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C22H26ClF2NO4 |
| Quantity | 50 mg |
| Synonym | R 065824 hydrochloride |
| Solubility Information | DMSO : 100 mg/mL (226.30 mM; Need ultrasonic) ∣H2O : 1.79 mg/mL (4.05 mM; ultrasonic and warming and heat to 60°C) |
| SMILES | FC1=CC=C2C(CC[C@]([C@@H](O)CNC[C@H](O)[C@@]3([H])CCC(C=C(F)C=C4)=C4O3)([H])O2)=C1.Cl |
| Molecular Weight (g/mol) | 441.9 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction