missing translation for 'onlineSavingsMsg'
Learn More

Memmert™ Nearly Gastight Interior with Gas Inlet/Outlet for UN110

Product Code. 17529785 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UN110 Oven
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17529785

Brand: Memmert™ UN110K3

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Nearly gastight interior with possibility for inlet/outlet through two tubes with ball valves.

  • Gas inlet/outlet through 2 tubes with ball valves
  • Technical clarification required for combination with other options

Specifications

For Use With (Equipment) UN110 Oven
Type Interior
Includes Gas Inlet/Outlet
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.