Learn More
Memmert™ Nearly Gastight Interior with Gas Inlet/Outlet for UF260
Product Code. 17519155 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UF260 Oven
Unit Size:
Each
Product Code. 17519155
Brand: Memmert™ UF260K3
Only null left
Add to basket
Add to basket
Nearly gastight interior with possibility for inlet/outlet through two tubes with ball valves.
  • Gas inlet/outlet through 2 tubes with ball valves
  • Technical clarification required for combination with other options
Specifications
For Use With (Equipment) UF260 Oven
Type Interior
Includes Gas Inlet/Outlet