Learn More
Memmert™ Nearly Gastight Interior with Gas Inlet/Outlet for UF160plus
Product Code. 17539095 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UF160plus Oven
Unit Size:
Each
Product Code. 17539095
Brand: Memmert™ UF160plusK3
Only null left
Add to basket
Add to basket
Nearly gastight interior with possibility for inlet/outlet through two tubes with ball valves.
  • Gas inlet/outlet through 2 tubes with ball valves
  • Technical clarification required for combination with other options
Specifications
For Use With (Equipment) UF160plus Oven
Type Interior
Includes Gas Inlet/Outlet