missing translation for 'onlineSavingsMsg'
Learn More

Neamine, TRC

Product Code. 17994495
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17994495 10 mg 10mg
17954335 25 mg 25mg
17952665 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17994495 Supplier Toronto Research Chemicals Supplier No. TRCN38601010MG
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3947-65-7
Molecular Formula C12 H26 N4 O6
Molecular Weight (g/mol) 322.36
Synonym 2-Deoxy-4-O-(2,6-diamino-2,6-dideoxy-α-D-glucopyranosyl)-D-streptamine, Neomycin A (6CI,7CI,8CI), (+)-Neamine, Dekamycin V, Neamin, Neamine, Nebramycin X, Negamicin, O-2,6-Diamino-2,6-dideoxy-α-D-glucopyranosyl-(1' ' ’4)-1,3-diamino-1,2,3-trideoxy-D-myo-inositol, ST 7, Neomycin Imp. A (EP), GW 318764X
IUPAC Name (2R,3S,4R,5R,6R)-5-amino-2-(aminomethyl)-6-[(1R,2R,3S,4R,6S)-4,6-diamino-2,3-dihydroxycyclohexyl]oxyoxane-3,4-diol
SMILES NC[C@H]1O[C@H](O[C@@H]2[C@@H](N)C[C@@H](N)[C@H](O)[C@H]2O)[C@H](N)[C@@H](O)[C@@H]1O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C12H26N4O6/c13-2-5-8(18)9(19)6(16)12(21-5)22-11-4(15)1-3(14)7(17)10(11)20/h3-12,17-20H,1-2,13-16H2/t3-,4+,5-,6-,7+,8-,9-,10-,11-,12-/m1/s1
Formula Weight 322.1852
Percent Purity >95%
Chemical Name or Material Neamine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.