Learn More
HUBER Silicone Hose, Pressureless, S-180-NW8
Suitable hose for pressureless applications and cooling water. HUBER; Silicone Hose, Pressureless, S-180-NW8 is not suitable for thermofluids that contain silicone.
Brand: HUBER
Code : 04
Additional Details : Weight : 0.06000kg
Description
- Specifically designed for use in pressureless applications and with cooling water.
- Silicone construction.
- Thermally resistant.
- Not for use with thermofluids that contain silicone.
Specifications
Silicone Rubber |